Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erk1/2, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89138062 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-138-062 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-138-062 Supplier Abnova Corporation Supplier No. PAB15417
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Erk1/2.

Specifications

Antigen Erk1/2
Applications Flow Cytometry, Immunohistochemistry, Western Blot, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Erk1/2.
Dilution Flow Cytometry (10 ug/mL) Immunohistochemistry (1:50) Western Blot (1:5000) The optimal working dilution should be determined by the end user.
Formulation In serum
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to 35 amino acids of Erk1/2 with the CGG spacer group added.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects ∼44KDa and ∼42KDa bands corresponding to the molecular weights of Erk1 and Erk2.
Target Species Bovine, Chicken, Frog, Fruitfly, Human, Mouse, Rat, Sheep, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.