Learn More
Description
Specifically detects ERLIN1/SPFH1 in Mouse, Rat samples, and it is validated for Immunocytochemistry, Immunoprecipitation, Western Blotting
Specifications
Specifications
| Antigen | ERLIN1/SPFH1 |
| Applications | Immunocytochemistry, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Formulation | Purified rabbit polyclonal in buffer containing 155mM NaCl, 50mM Tris-HCl pH 7.4, with 0.05% sodium azide and 10% glycerol. |
| Gene Accession No. | B1WBY7 |
| Gene Symbols | Erlin1; Spfh1_predicted; rCG_57742 |
| Host Species | Rabbit |
| Immunogen | Linear peptide corresponding to the C-terminus of rat ERLIN1/SPFH1. |
| Purification Method | Affinity Purified |
| Quantity | 200 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.