Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ESRRA Rabbit anti-Bovine, Dog, Hamster, Horse, Human, Monkey, Mouse, Pig, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89129295 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-129-295 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-129-295 Supplier Abnova Corporation Supplier No. PAB27776
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of ESRRA.

The protein encoded by this gene is a nuclear receptor that is closely related to the estrogen receptor. This protein acts as a site-specific transcription regulator and has been also shown to interact with estrogen and the transcripton factor TFIIB by direct protein-protein contact. The binding and regulatory activities of this protein have been demonstrated in the regulation of a variety of genes including lactoferrin, osteopontin, medium-chain acyl coenzyme A dehydrogenase (MCAD) and thyroid hormone receptor genes. A processed pseudogene of ESRRA is located on chromosome 13q12.1. [provided by RefSeq

Specifications

Antigen ESRRA
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of ESRRA.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (11-21 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene ESRRA
Gene Accession No. P11474
Gene Alias ERR1/ERRa/ERRalpha/ESRL1/NR3B1
Gene Symbols ESRRA
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 15 amino acid at ligand-binding domain of human ESRRA.
Purification Method Immunoaffinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2101
Test Specificity BLAST analysis of the peptide immunogen showed no homology with other human proteins, except ESRRG (60%).
Target Species Bovine, Canine, Equine, Hamster, Human, Monkey, Mouse, Pig, Rat
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.