Learn More
ETNK1 Rabbit anti-Hamster, Human, Mouse, Polyclonal Antibody, Abnova™
Catalog No. 17720145
Catalog No. 17-720-145 Supplier Thermo Scientific Supplier No. 82000125RX
Add to Cart
Add to Cart
Request a Quote
Rabbit polyclonal antibody raised against synthetic peptide of ETNK1.
This gene encodes an ethanolamine kinase, which functions in the first committed step of the phosphatidylethanolamine synthesis pathway. This cytosolic enzyme is specific for ethanolamine and exhibits negligible kinase activity on choline. Alternative splicing results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq