Learn More
ETNK1 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ETNK1.
Supplier: Abnova Corporation PAB4608
Description
This gene encodes an ethanolamine kinase, which functions in the first committed step of the phosphatidylethanolamine synthesis pathway. This cytosolic enzyme is specific for ethanolamine and exhibits negligible kinase activity on choline. Alternative splicing results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq
Specifications
ETNK1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of ETNK1. | |
In PBS (0.09% sodium azide) | |
Q9HBU6 | |
ETNK1 | |
A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human ETNK1. | |
400 μL | |
Primary | |
Human, Mouse | |
Liquid |
ELISA, Western Blot | |
Unconjugated | |
ELISA (1:1000) Western Blot (1:100-500) The optimal working dilution should be determined by the end user. | |
ETNK1 | |
EKI/EKI1/Nbla10396 | |
Rabbit | |
Protein G purification | |
RUO | |
55500 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only