Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EXOC8, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89123604 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-123-604 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-123-604 Supplier Abnova Corporation Supplier No. PAB21899
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant EXOC8.

Sequence: RDHLRGQAGFFSTPGGASRDGSGPGEEGKQRTLTTLLEKVEGCRHLLETPGQYLVYNGDLVEYDADHMAQLQRVHGFLMNDCLLVATWLPQRRGMYRYNAL

Specifications

Antigen EXOC8
Applications Immunofluorescence, Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant EXOC8.
Dilution Immunohistochemistry (1:200-1:500) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene EXOC8
Gene Accession No. Q8IYI6
Gene Alias EXO84/Exo84p/SEC84
Gene Symbols EXOC8
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human EXOC8.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 149371
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.