Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-Exportin-1/CRM1, Polyclonal,

Catalog No. ABS1626MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS1626MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS1626MI Supplier MilliporeSigma™ Supplier No. ABS1626
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Exportin-1/CRM1 in Human, Monkey, Mouse samples, and it is validated for Immunoprecipitation, Western Blotting

Exportin-1 (UniProt O14980; also known as Chromosome region maintenance 1 protein homolog, CRM1 homolog, Exp1) is encoded by the XPO1 (also known as CRM1) gene (Gene ID 7514) in human. The transport of macromolecules between cytoplasm and nucleus is mediated by importins and exportins and regulated by the small GTPase Ran. Most nuclear importins & exportins, including exportin-1, belong to the beta-karyopherin (KAP) family of proteins, Importins bind their cargoes with either nuclear localization signal (NLS) or M9 domain-containing hnRNP proteins in the cytoplasm, while exportins target their substrates via their nuclear export signal (NES) or M9 domain inside the nucleus. The energy dependence of nucleocytoplasmic transport replies on the Ran GTPase system, where the nucleotide exchange factor (NEF) RCC1/Prp20p, the GTPase activating protein (GAP) RanGAP1/Rna1p, and the Ran binding protein (RBP) RanBP1/Yrb1p are arranged asymmetrically to produce a high Ran GTP:Ran GDP ratio in the nucleus and the reverse ratio in the cytoplasm. Exportin 1/CRM1 is reported to bind RanGTP and either Rev or PKI NES peptides in a cooperative manner in vitro and exogenous expression of human CRM1 in Xenopus oocytes is shown to increase the rate of Rev and U snRNA nuclear export.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Exportin-1/CRM1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Formulation Rabbit polyclonal serum with 0.05% sodium azide.
Gene Accession No. O14980
Gene Symbols XPO1; CRM1
Host Species Rabbit
Immunogen Recombinant protein corresponding to the C-terminus of human Exportin-1/CRM1.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_003391
Target Species Human, Monkey, Mouse
Content And Storage Stable for one year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.