Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EYA4 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89125041 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-125-041 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-125-041 Supplier Abnova Corporation Supplier No. PAB23343
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant EYA4.

This gene encodes a member of the eyes absent (EYA) family of proteins. The encoded protein may act as a transcriptional activator through its protein phosphatase activity, and it may be important for eye development, and for continued function of the mature organ of Corti. Mutations in this gene are associated with postlingual, progressive, autosomal dominant hearing loss at the deafness, autosomal dominant nonsyndromic sensorineural 10 locus. Defects in this gene are also associated with dilated cardiomyopathy 1J. Three transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq

Sequence: TGGENMTVLNTADWLLSCNTPSSATMSLLAVKTEPLNSSETTATTGDGALDTFTGSVITSSGYSPRSA

Specifications

Antigen EYA4
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant EYA4.
Dilution Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene EYA4
Gene Accession No. O95677
Gene Alias CMD1J/DFNA10
Gene Symbols EYA4
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human EYA4.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2070
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.