Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EZH1 Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89121801 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-121-801 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-121-801 Supplier Abnova Corporation Supplier No. PAB20297

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit polyclonal antibody raised against recombinant EZH1.

EZH1 is a component of a noncanonical Polycomb repressive complex-2 (PRC2) that mediates methylation of histone H3 (see MIM 602812) lys27 (H3K27) and functions in the maintenance of embryonic stem cell pluripotency and plasticity (Shen et al., 2008 [PubMed 19026780]).[supplied by OMIM

Sequence: MGDEVKEEDETFIEELINNYDGKVHGEEEMIPGSVLISDAVFLELVDALNQYSDEEEEGHNDTSDGKQDDSKEDLPVTRKRKRHAIEGNKKSSKKQFPNDMIFSAIASMFPENGVPDDMKERYRELTEMSDPNALPPQCTPNIDGPNA

Specifications

Antigen EZH1
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Description enhancer of zeste homolog 1 (Drosophila)
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene EZH1
Gene Accession No. Q92800
Gene Alias KIAA0388
Gene Symbols EZH1
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human EZH1.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2145
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.