Learn More
EZH2 Rabbit anti-Human, Mouse, Primates, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of EZH2.
Supplier: Abnova Corporation PAB12430
Description
This gene encodes a member of the Polycomb-group (PcG) family. PcG family members form multimeric protein complexes, which are involved in maintaining the transcriptional repressive state of genes over successive cell generations. This protein associates with the embryonic ectoderm development protein, the VAV1 oncoprotein, and the X-linked nuclear protein. This protein may play a role in the hematopoietic and central nervous systems. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq
Specifications
EZH2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of EZH2. | |
In Tris-citrate/phosphate buffer, pH 7.0-8.0 (0.09% sodium azide) | |
Q15910 | |
EZH2 | |
A synthetic peptide corresponding to amino acids 300-400 of human EZH2. | |
RUO | |
2146 | |
Store at 4°C. Do not freeze. |
Western Blot | |
Unconjugated | |
Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
EZH2 | |
ENX-1/EZH1/KMT6/MGC9169 | |
Rabbit | |
100 μL | |
Primary | |
Human, Mouse, Primate | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only