Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

F13A1/F13A2/F13A3/F13B, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89121388 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
150 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-121-388 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-121-388 Supplier Abnova Corporation Supplier No. PAB19873
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against native F13A1/F13A2/F13A3/F13B.

Specifications

Antigen F13A1/F13A2/F13A3/F13B
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against native F13A1/F13A2/F13A3/F13B.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (0.02% sodium azide, 50% glycerol (v/v))
Gene Alias F13A
Gene Symbols F13A1
Host Species Rabbit
Immunogen Native purified F13A1/F13A2/F13A3/F13B from human plasma.
Purification Method Affinity purification
Quantity 150 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2162/2163/2164/2165
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.