Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

F2 Rabbit anti-Pig, Polyclonal Antibody, Abnova™

Catalog No. 89121407 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
150 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-121-407 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-121-407 Supplier Abnova Corporation Supplier No. PAB19894
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against native F2.

Specifications

Antigen F2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against native F2.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (0.02% sodium azide, 50% glycerol (v/v))
Gene F2
Gene Symbols F2
Host Species Rabbit
Immunogen Native purified F2 from porcine plasma.
Purification Method Affinity purification
Quantity 150 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100144442
Target Species Pig
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.