Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
F2RL1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of F2RL1.
Supplier: Abnova Corporation PAB24714
Description
Sequence: YFVSKDFRDHARNALLCSpecifications
F2RL1 | |
Polyclonal | |
Unconjugated | |
WB (1:250) Immunoprecipitation (1:200) The optimal working dilution should be determined by the end user. | |
F2rl1 | |
Gpcr11/PAR-2/Par2 | |
Rabbit | |
Affinity Purification | |
RUO | |
14063 | |
Store at -20°C Aliquot to avoid repeated freezing and thawing. |
Immunoprecipitation, Western Blot | |
0.6-0.65 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of F2RL1. | |
In Tris-glycine buffer, pH 7.4 (stabilizing proteins, cryogenic stabilizers, glycerol, 0.02% sodium azide) | |
NP_032000 | |
F2rl1 | |
A synthetic peptide corresponding to amino acids 347-363 of mouse F2rl1. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction