Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

F2rl2, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89126380 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-126-380 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-126-380 Supplier Abnova Corporation Supplier No. PAB24715
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of F2rl2.

Sequence: KPTLTIKSFNGGPQNTFEEFPLSDIE

Specifications

Antigen F2rl2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.51-0.54 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of F2rl2.
Dilution WB (1:250) Immunoprecipitation (1:200) The optimal working dilution should be determined by the end user.
Formulation In Tris-glycine buffer, pH 7.4 (stabilizing proteins, cryogenic stabilizers, glycerol, 0.02% sodium azide)
Gene F2rl2
Gene Accession No. NP_034300
Gene Alias F730031A08/PAR3
Gene Symbols F2rl2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 31-355 of mouse F2rl2.
Purification Method Affinity Purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14064
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.