Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-FABP7, Polyclonal,

Catalog No. ABS811MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS811MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS811MI Supplier MilliporeSigma™ Supplier No. ABS811
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects FABP7 in Human samples, and it is validated for Immunohistochemistry, Western Blotting

FABP7 is a member of the large family of hydrophobic ligand-binding proteins (FABPs). FABP proteins are involved in lipid metabolism, including uptake and intracellular trafficking of fatty acids and retinoids. In addition, they play a role in gene regulation, cell signaling, cell growth and differentiation. Several reports have suggested a possible role for the different FABP proteins in cancer biology, linking their levels with either increasing or decreasing the degree of malignancy. Nine FABPs (FABP1-FABP9) have been identified. FABP7 is a brain-specific protein that is required for the establishment of the radial glial fiber system in the developing brain, a system that is necessary for the migration of immature neurons to establish cortical layers. It is expressed at high levels in the developing central nervous system. FABP7 has also been found in melanocytic lesions and has shown to regulate proliferation and invasion in melanoma cells in vitro, suggesting that it may play a role in cell proliferation and invasion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FABP7
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Formulation Purified rabbit polyclonal in buffer containing PBS (pH 7.2) with 0.02% sodium azide.
Gene Accession No. O15540
Gene Symbols FABP7; BLBP; FABPB; MRG
Host Species Rabbit
Immunogen FABP7 antibody was raised against a 17 amino acid synthetic peptide from near the center of human FABP7.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.