Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Factor VIIa, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89136881 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-881 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-881 Supplier Abnova Corporation Supplier No. PAB0124
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant Factor VIIa.

Specifications

Antigen Factor VIIa
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant Factor VIIa.
Dilution ELISA (1:15000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS, pH 7.4
Host Species Rabbit
Immunogen Recombinant human Factor VIIa.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at -20°C on dry atmosphere.
Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.