Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM167B, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant FAM167B.
Supplier: Abnova Corporation PAB22281
Description
Sequence: MQAQDRQLAGQLLRLRAQLHRLKMDQACHLHQELLDEAELELELEPGAGLALAPLLRHLGLTRMNISARRFTLCSpecifications
FAM167B | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant FAM167B. | |
In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
Q9BTA0 | |
FAM167B | |
Recombinant protein corresponding to amino acids of human FAM167B. | |
100 μL | |
Primary | |
Human | |
Liquid |
Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunohistochemistry (1:20-1:50) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
FAM167B | |
C1orf90/MGC10820 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
84734 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction