Learn More
FAM219A, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant FAM219A.
Supplier: Abnova Corporation PAB21199
Description
The protein encoded by this gene has homologs that have been identified in mouse and macaque. The mouse and human proteins have a putative prenyl group binding site (CAAX box) at their C-terminus. A diverse list of proteins are known or strongly presumed to be the target of post-translational modification by the attachment of either a farnesyl or a geranyl-geranyl group to a cysteine residue at the C-terminus. The function of this protein has not been determined. [provided by RefSeq
Sequence: MEEIDRFQVPTAHSEMQPLDPAAASISDGDCDAREGESVAMNYKPSPLQVKLEKQRELARKGSLKNGSMSpecifications
FAM219A | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant FAM219A. | |
In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
Q8IW50 | |
FAM219A | |
Recombinant protein corresponding to amino acids of human FAM219A. | |
100 μL | |
Primary | |
Human | |
Liquid |
Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user. | |
FAM219A | |
RP11-573M23.5/C9orf25/bA573M23.5 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
203259 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only