Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM71F1 Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89122892 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-122-892 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-122-892 Supplier Abnova Corporation Supplier No. PAB21401

Rabbit polyclonal antibody raised against recombinant FAM71F1.

Sequence: GLSITTKDPRILVTHCLVPKNCSSPSGDSKLVQKKLQASQPSESLIQLMTKGESEALSQIFADLHQQNQ

Specifications

Antigen FAM71F1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description family with sequence similarity 71, member F1
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene FAM71F1
Gene Accession No. Q96KD3
Gene Alias FAM137A, NYD-SP18
Gene Symbols FAM71F1
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human FAM71F1.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84691
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.