Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-FATP4, Polyclonal,

Catalog No. ABN1036MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN1036MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN1036MI Supplier MilliporeSigma™ Supplier No. ABN1036
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects FATP4 in Human samples, and it is validated for Immunocytochemistry, Immunohistochemistry

Fatty acid transport proteins (FATP) are multifunctional proteins that are involved in the translocation of long-chain fatty acids across the plasma membrane. FATP4 is the only member of this family that is expressed in the small intestine and is localized to the apical brush border of the epithelial cells, where it is responsible for the uptake of dietary lipids. FATP4 exhibits acyl-CoA ligase activity for long-chain and very-long chain fatty acids. Its expression is high in testis, brain, and kidney and very low levels are found in peripheral leukocytes, bone marrow, and skeletal muscle. Defects and depletion of FATP4 have been linked to abnormalities in skin homeostasis and altered insulin sensitivity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FATP4
Applications Immunocytochemistry, Immunohistochemistry
Classification Polyclonal
Formulation Purified rabbit polyclonal IgG in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q6P1M0
Gene Symbols SLC27A4; ACSVL4; FATP4
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human FATP4 near the C-terminal.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_005085
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.