Learn More
FAU Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of FAU.
Supplier: Abnova Corporation PAB1712
Description
This gene is the cellular homolog of the fox sequence in the Finkel-Biskis-Reilly murine sarcoma virus (FBR-MuSV). It encodes a fusion protein consisting of the ubiquitin-like protein fubi at the N terminus and ribosomal protein S30 at the C terminus. It has been proposed that the fusion protein is post-translationally processed to generate free fubi and free ribosomal protein S30. Fubi is a member of the ubiquitin family, and ribosomal protein S30 belongs to the S30E family of ribosomal proteins. Whereas the function of fubi is currently unknown, ribosomal protein S30 is a component of the 40S subunit of the cytoplasmic ribosome. Pseudogenes derived from this gene are present in the genome. Similar to ribosomal protein S30, ribosomal proteins S27a and L40 are synthesized as fusion proteins with ubiquitin. [provided by RefSeq
Specifications
FAU | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of FAU. | |
In PBS (0.09% sodium azide) | |
NP_001988 | |
FAU | |
A synthetic peptide (conjugated with KLH) corresponding to internal region of human FAU. | |
400 μL | |
Primary | |
Human | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Western Blot (1:1000) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
FAU | |
FAU1/FLJ22986/Fub1/Fubi/MNSFbeta/RPS30 | |
Rabbit | |
Protein G purification | |
RUO | |
2197 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only