Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXO11, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89130194 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-194 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-194 Supplier Abnova Corporation Supplier No. PAB28680
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant FBXO11.

This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. Alternatively spliced transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]

Sequence: GHRAKRARVSGKSQDLSAAPAEQYLQEKLPDEVVLKIFSYLLEQDLCRAACVCKRFSELANDPILWKRLYMEVFEYTRPMMHPEPGKFYQINPEEYEHPNPWKESFQQLYKGAHVKPGFAEHFYSNPARYKGR

Specifications

Antigen FBXO11
Applications Immunofluorescence, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant FBXO11.
Dilution Immunohistochemistry (1:50-1:200) Immunofluorescence (1-4 ug/ml) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide).
Gene FBXO11
Gene Alias FBX11/FLJ12673/MGC44383/PRMT9/UBR6/UG063H01/VIT1
Gene Symbols FBXO11
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human FBXO11.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80204
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.