Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXO9, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89114554 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-554 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-554 Supplier Abnova Corporation Supplier No. PAB10012
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of FBXO9.

This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. Alternative splicing of this gene generates at least 3 transcript variants diverging at the 5' terminus. [provided by RefSeq]

Specifications

Antigen FBXO9
Applications ELISA, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of FBXO9.
Dilution ELISA (1:5000-1:25000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene FBXO9
Gene Accession No. NP_036479(isoform1);NP_258441(isoform2);NP_258442(isoform3)
Gene Alias DKFZp434C0118/FBX9/KIAA0936/NY-REN-57/VCIA1/dJ341E18.2
Gene Symbols FBXO9
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 431-447 of human FBXO9.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26268
Test Specificity Reactivity is expected against all known isoforms of this protein, as the immunogen sequence is common to their carboxy-terminal ends.
Target Species Bovine, Canine, Chicken, Chimpanzee, Frog, Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.