Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-FcRn/FCGRT, Polyclonal,

Catalog No. ABF189MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABF189MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABF189MI Supplier MilliporeSigma™ Supplier No. ABF189
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects FcRn/FCGRT in Human samples, and it is validated for Western Blotting

IgG receptor FcRn large subunit p51 (UniProt P55899; also known as FcRn, IgG Fc fragment receptor transporter alpha chain, Neonatal Fc receptor) is encoded by the FCGRT gene (also known as FCRN) (Gene ID 2217) in human. Transfer of passive immunity from mother to the fetus or newborn involves the transport of IgG across several epithelia. Depending on the species, IgG is transported prenatally across the placenta and yolk sac or is absorbed from colostrum and milk by the small intestine of the suckling newborn. In both cases, apical to basolateral transepithelial transport of IgG is mediated by FcRn. FcRn is a heterodimer of an alpha-chain and beta(2)-microglobulin (beta(2)m) and differs from other IgG Fc receptors in that it is structurally related to MHC class I molecules. The assembly of the FcRn alpha-chain with beta(2)m is important for both cell surface expression and pH-dependent high affinity IgG binding. Proteins endocytosed from serum are degraded in the lysosomes. However, serum albumin (SA) and IgG bind to FcRn at low pH in the endosome after endocytosis, and are transported back to the cellular surface, where they are released into the bloodstream, resulting in an extended serum half-life.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FcRn/FCGRT
Applications Western Blot
Classification Polyclonal
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P55899
Gene Symbols FCGRT; FCRN
Host Species Rabbit
Immunogen Linear peptide corresponding to the Alpha-2/-3 domain of human FcRn/FCGRT.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_001129491
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.