Learn More
FFAR1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of FFAR1.
Supplier: Abnova Corporation PAB26307
Description
This gene encodes a member of the GP40 family of G protein-coupled receptors that are clustered together on chromosome 19. The encoded protein is a receptor for medium and long chain free fatty acids and may be involved in the metabolic regulation of insulin secretion. Polymorphisms in this gene may be associated with type 2 diabetes. [provided by RefSeq
Specifications
FFAR1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of FFAR1. | |
In PBS (0.09% sodium azide) | |
O14842 | |
FFAR1 | |
A synthetic peptide corresponding to 18 amino acids at C-terminus of human FFAR1. | |
50 μg | |
Primary | |
BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (9-18 ug/mL) The optimal working dilution should be determined by the end user. | |
FFAR1 | |
FFA1R/GPCR40/GPR40 | |
Rabbit | |
Immunoaffinity chromatography | |
RUO | |
2864 | |
Human, Monkey | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only