Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGG, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89117342 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-342 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-342 Supplier Abnova Corporation Supplier No. PAB14903
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against FGG.

The protein encoded by this gene is the gamma component of fibrinogen, a blood-borne glycoprotein comprised of three pairs of nonidentical polypeptide chains. Following vascular injury, fibrinogen is cleaved by thrombin to form fibrin which is the most abundant component of blood clots. In addition, various cleavage products of fibrinogen and fibrin regulate cell adhesion and spreading, display vasoconstrictor and chemotactic activities, and are mitogens for several cell types. Mutations in this gene lead to several disorders, including dysfibrinogenemia, hypofibrinogenemia and thrombophilia. Alternative splicing results in two transcript variants encoding different isoforms. [provided by RefSeq

Specifications

Antigen FGG
Applications ELISA, Immunocytochemistry, Immunodiffusion, Immunomicroscopy, Radioimmune Assays (RIA)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against FGG.
Dilution ELISA (1:10000) Immunodiffusion (1:32) The optimal working dilution should be determined by the end user.
Formulation In serum
Gene FGG
Gene Symbols FGG
Host Species Rabbit
Immunogen Human FGG.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2266
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.