Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FHOD3 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89123440 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-123-440 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-123-440 Supplier Abnova Corporation Supplier No. PAB21739
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant FHOD3.

Proteins that contain formin (FMN; MIM 136535) homology (FH) domains, such as FHOD3, play a role in regulation of the actin cytoskeleton (Kanaya et al., 2005 [PubMed 15966898]).[supplied by OMIM

Sequence: QCGDILTNKRFMLDMLYAHNRKSPDDEEKGDGEAGRTQQEAEAVASLATRISTLQANSQTQDESVRRVDVGCLDNRGSVKAFAEKFNSGDLGRGSISPDAEPNDKVPETAPVQPKTESDYIWDQLM

Specifications

Antigen FHOD3
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant FHOD3.
Dilution Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene FHOD3
Gene Accession No. Q2V2M9
Gene Alias FHOS2/Formactin2
Gene Symbols FHOD3
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human FHOD3.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80206
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.