Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FITC, Rabbit, HRP, Polyclonal Antibody, Abnova™

Catalog No. 89137875 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-875 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-875 Supplier Abnova Corporation Supplier No. PAB14241
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against FITC.

Specifications

Antigen FITC
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate HRP
Description Rabbit polyclonal antibody raised against FITC.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (10mg/mL BSA, 0.002% thimerosal)
Host Species Rabbit
Immunogen FITC-KLH conjugate.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes Fluorescein. No cross-reactivity with immunoglobulin of any species.
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.