Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fkbp1a, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Fkbp1a.
Supplier: Abnova Corporation PAB8720
Specifications
Fkbp1a | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of Fkbp1a. | |
In PBS, pH 7.2 (30% glycerol, 0.5mg/mL BSA, 0.01% thimerosal) | |
12kDa/FKBP12/FKBP12-T1/FKBP12-T2/Fkbp/Fkbp1/mFKBP1/mFKBP12 | |
Rabbit | |
100 μg | |
Primary | |
This antibody detects 12 KDa FKBP12. | |
Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
Western Blot (0.5-4 ug/mL) Immunoprecipitation (2-10 ug/mL) Immunohistochemistry (2-10 ug/mL) The optimal working dilution should be determined by the end user. | |
Fkbp1a | |
Fkbp1a | |
A synthetic peptide corresponding to residues surrounding amino acids 10 of mouse Fkbp1a. | |
RUO | |
14225 | |
Bovine, Human, Mouse, Rabbit, Rat | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction