Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fkbp4, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89133880 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-133-880 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-133-880 Supplier Abnova Corporation Supplier No. PAB8722

Rabbit polyclonal antibody raised against synthetic peptide of Fkbp4.

Specifications

Antigen Fkbp4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description FK506 binding protein 4
Formulation In PBS, pH 7.2 (30% glycerol, 0.5mg/mL BSA, 0.01% thimerosal)
Gene Fkbp4
Gene Alias 59kDa, AL022792, AW208983, FKBP-52, FKPB52, p59
Gene Symbols Fkbp4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to residues surrounding amino acids 9 of mouse Fkbp4.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14228
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.