Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FLJ35848 Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89123061 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-123-061 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-123-061 Supplier Abnova Corporation Supplier No. PAB21565

Rabbit polyclonal antibody raised against recombinant FLJ35848.

Sequence: QQLGSNGFPLRSTHPFGHSVVPLLDSYDLLSYDDLSHLYPYFNMMYGDNSFSGLMPTFGFQRPIKTRSGPASELHIRLEECCEQWRALEKERKKTELALAKNYPGKKVSSTNNTPVPRLTSNPSRVDRLIVDELRELAR

Specifications

Antigen FLJ35848
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Description hypothetical protein FLJ35848
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene FLJ35848
Gene Accession No. A2RUB1
Gene Alias MGC43301
Gene Symbols FLJ35848
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human FLJ35848.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284071
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.