Learn More
FLT4 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of FLT4.
Supplier: Abnova Corporation PAB11653
Description
This gene encodes a tyrosine kinase receptor for vascular endothelial growth factors C and D. The protein is thought to be involved in lymphangiogenesis and maintenance of the lymphatic endothelium. Mutations in this gene cause hereditary lymphedema type IA. [provided by RefSeq
Specifications
FLT4 | |
Polyclonal | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (Formalin/PFA-fixed paraffin-embedded sections) (1:100) The optimal working dilution should be determined by the end user. | |
FLT4 | |
FLT4 | |
A synthetic peptide corresponding to C-terminus of human FLT4. | |
100 μL | |
Primary | |
Human | |
Liquid |
Immunohistochemistry (PFA fixed) | |
Lot specific | |
Rabbit polyclonal antibody raised against synthetic peptide of FLT4. | |
In 10mM PBS, pH 7.4 (1% BSA, 0.09% sodium azide) | |
FLT41/LMPH1A/PCL/VEGFR3 | |
Rabbit | |
Immunoaffinity purification | |
RUO | |
2324 | |
Store at 4°C. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only