Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FMNL3, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89130099 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-099 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-099 Supplier Abnova Corporation Supplier No. PAB28587
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant FMNL3.

The protein encoded by this gene contains a formin homology 2 domain and has high sequence identity to the mouse Wbp3 protein. Two alternative transcripts encoding different isoforms have been described. [provided by RefSeq

Sequence: LLDVKELGRGMELIRRECSIHDNSVLRNFLSTNEGKLDKLQRDAKTAEEAYNAVVRYFGESPKTTPPSVFFPVFVRFIRSYKEAEQENEARKKQEEVMREKQLAQEAKKLDAKTPSQRNKWQQQE

Specifications

Antigen FMNL3
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant FMNL3.
Dilution Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide)
Gene FMNL3
Gene Accession No. Q8IVF7
Gene Alias DKFZp762B245/FHOD3/FLJ45265/MGC45819/WBP3
Gene Symbols FMNL3
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human FMNL3.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91010
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.