Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FN1 Rabbit anti-Bovine, Polyclonal Antibody, Abnova™

Catalog No. 89116718 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-718 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-718 Supplier Abnova Corporation Supplier No. PAB13517
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against native FN1.

Specifications

Antigen FN1
Applications ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Radioimmune Assays (RIA)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against native FN1.
Dilution ELISA (1:200) Immunohistochemistry (1:500) Immunofluorescence (1:80) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from phosphate buffer
Gene FN1
Gene Symbols FN1
Host Species Rabbit
Immunogen Native purified FN1 from bovine plasma.
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 280794
Test Specificity This antibody reacts with bovine fibronectin 100%, bovine collagen type I and III <0.1% (RIA at 1:10000 dilution).
Target Species Bovine
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with 0.5mL distilled water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.