Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fn1 Rabbit anti-Rat, Polyclonal Antibody, Abnova™

Catalog No. 89116720 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-720 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-720 Supplier Abnova Corporation Supplier No. PAB13519

Rabbit polyclonal antibody raised against native Fn1.

Specifications

Antigen Fn1
Applications ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Radioimmune Assays (RIA), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against native Fn1.
Dilution ELISA (1:200) Immunohistochemistry (1:1000) Immunofluorescence (1:80) The optimal working dilution should be determined by the end user.
Formulation In phosphate buffer
Gene Fn1
Gene Alias FIBNEC/fn-1
Gene Symbols Fn1
Host Species Rabbit
Immunogen Native purified Fn1 from rat plasma.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25661
Test Specificity This antibody reacts with rat fibronectin 100%, rat collagen type I, II, III, and V <0.1%, rat elastin <0.1% (RIA at 1:5000 dilution).
Target Species Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.