Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FNIP1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89115747 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-747 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-747 Supplier Abnova Corporation Supplier No. PAB12367
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of FNIP1.

Sequence: MAPTLFQKLFSKRTGLGAC

Specifications

Antigen FNIP1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of FNIP1.
Dilution Western Blot (1:1000-1:2000) The optimal working dilution should be determined by the end user.
Formulation In antiserum (0.09% sodium azide)
Gene FNIP1
Gene Alias DKFZp686E18167/DKFZp781P0215/KIAA1961/MGC667
Gene Symbols FNIP1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to human FNIP1.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 96459
Test Specificity human and mouse FNIP1.
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.