Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FNTA Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89131367 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-131-367 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-131-367 Supplier Abnova Corporation Supplier No. PAB4326
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of FNTA.

Prenyltransferases attach either a farnesyl group or a geranylgeranyl group in thioether linkage to the cysteine residue of protein's with a C-terminal CAAX box. CAAX geranylgeranyltransferase and CAAX farnesyltransferase are heterodimers that share the same alpha subunit but have different beta subunits. This gene encodes the alpha subunit of these transferases. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq]

Specifications

Antigen FNTA
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of FNTA.
Dilution ELISA (1:1000) Western Blot (1:100-500) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene FNTA
Gene Accession No. NP_002018;P49354
Gene Alias FPTA/MGC99680/PGGT1A/PTAR2
Gene Symbols FNTA
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to internal region of human FNTA.
Purification Method Protein G purification
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2339
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.