Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FNTA Rabbit anti-Chicken, Chimpanzee, Dog, Human, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89114122 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-122 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-122 Supplier Abnova Corporation Supplier No. PAB0301
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant FNTA.

Prenyltransferases attach either a farnesyl group or a geranylgeranyl group in thioether linkage to the cysteine residue of protein's with a C-terminal CAAX box. CAAX geranylgeranyltransferase and CAAX farnesyltransferase are heterodimers that share the same alpha subunit but have different beta subunits. This gene encodes the alpha subunit of these transferases. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq]

Specifications

Antigen FNTA
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant FNTA.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% gelatin, 0.05% sodium azide)
Gene FNTA
Gene Accession No. NP_002018
Gene Alias FPTA/MGC99680/PGGT1A/PTAR2
Gene Symbols FNTA
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 150-250 of human FNTA.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2339
Target Species Canine, Chicken, Chimpanzee, Human, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.