Learn More
FOS (phospho T232), Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of FOS.
Supplier: Abnova Corporation PAB0475
Description
The Fos gene family consists of 4 members: FOS, FOSB, FOSL1, and FOSL2. These genes encode leucine zipper proteins that can dimerize with proteins of the JUN family, thereby forming the transcription factor complex AP-1. As such, the FOS proteins have been implicated as regulators of cell proliferation, differentiation, and transformation. In some cases, expression of the FOS gene has also been associated with apoptotic cell death. [provided by RefSeq
Specifications
FOS | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of FOS. | |
In PBS (0.09% sodium azide) | |
NP_005243;P01100 | |
FOS | |
Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding T232 of human FOS. | |
400 μL | |
Primary | |
Human | |
Liquid |
Dot Blot, ELISA | |
Unconjugated | |
Dot Blot (1:100-500) The optimal working dilution should be determined by the end user. | |
FOS | |
AP-1/C-FOS | |
Rabbit | |
Protein A purification | |
RUO | |
2353 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only