Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOS (phospho T232), Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89130659 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-659 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-659 Supplier Abnova Corporation Supplier No. PAB29224
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic phosphopeptide of human FOS.

The Fos gene family consists of 4 members: FOS, FOSB, FOSL1, and FOSL2. These genes encode leucine zipper proteins that can dimerize with proteins of the JUN family, thereby forming the transcription factor complex AP-1. As such, the FOS proteins have been implicated as regulators of cell proliferation, differentiation, and transformation. In some cases, expression of the FOS gene has also been associated with apoptotic cell death. [provided by RefSeq

Specifications

Antigen FOS
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic phosphopeptide of human FOS.
Dilution Immunohistochemistry (1:50-1:100) Western Blot (1:500-1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (without mg2+ and Ca2+), (50% glycerol, 0.02% sodium azide)
Gene FOS
Gene Accession No. P01100
Gene Alias AP-1/C-FOS
Gene Symbols FOS
Host Species Rabbit
Immunogen Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding T232 of human FOS.
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2353
Test Specificity This antibody detects endogenous levels of FOS only when phosphorylated at threonine 232.
Target Species Human, Mouse
Content And Storage Store at -20°C.
Avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.