Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-FOXK2, Polyclonal,

Catalog No. ABE1828MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE1828MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE1828MI Supplier MilliporeSigma™ Supplier No. ABE1828
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects FOXK2 in Human samples, and it is validated for Immunoprecipitation, Western Blotting

Forkhead box protein K2 (UniProt Q01167; also known as FOXK2, Cellular transcription factor ILF-1, Interleukin enhancer-binding factor 1) is encoded by the FOXK2 (also known as ILF, ILF-1, ILF1) gene (Gene ID 3607) in human. FOXK2 is a transcription factor involved in cell proliferation and survival. FOXK1 and FOXK2 belong to the family of Fox transcription factors characterized by the forkhead DNA-binding domains. However, FOXK1 and FOXK2 can be distinguished from other Fox transcription factors by the presence of the forkhead-associated (FHA) domains. FOXK2 interacts with BRCA1-associated protein 1 (BAP1) through the FHA domain in a BAP1 Thr493 phosphorylation-dependent manner. BAP1 in turn recruits HCF-1, thereby forming a ternary complex at the FOXK2 target genes sites, where BAP1 represses FOXK2 target genes expression in a BAP1 DUB activity-dependent manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXK2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Formulation Rabbit polyclonal serum with 0.05% sodium azide.
Gene Accession No. Q01167
Gene Symbols FOXK2; ILF; ILF1
Host Species Rabbit
Immunogen Recombinant protein corresponding to the C-terminal region of human FOXK2.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_004505
Target Species Human
Content And Storage Stable for one year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.