Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXO4 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89133730 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-133-730 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-133-730 Supplier Abnova Corporation Supplier No. PAB8532

Rabbit polyclonal antibody raised against synthetic peptide of FOXO4.

Specifications

Antigen FOXO4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description forkhead box O4
Formulation In PBS, pH 7.2 (30% glycerol, 0.5mg/mL BSA, 0.01% thimerosal)
Gene FOXO4
Gene Alias AFX, AFX1, MGC120490, MLLT7
Gene Symbols FOXO4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to residues surrounding amino acids 491 of human FOXO4.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4303
Test Specificity This antibody detects 60kDa FOXO4 from samples of human, mouse, and rat origins.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.