Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXP3 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89115799 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-799 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-799 Supplier Abnova Corporation Supplier No. PAB12426
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of FOXP3.

The protein encoded by this gene is a member of the forkhead/winged-helix family of transcriptional regulators. Defects in this gene are the cause of immunodeficiency polyendocrinopathy, enteropathy, X-linked syndrome (IPEX), also known as X-linked autoimmunity-immunodeficiency syndrome. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq

Specifications

Antigen FOXP3
Applications Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of FOXP3.
Dilution Western Blot (1:1000-1:5000) Immunohistochemistry (Frozen sections) (1:50) The optimal working dilution should be determined by the end user.
Formulation In Tris-citrate/phosphate buffer, pH 7.0-8.0
Gene FOXP3
Gene Accession No. Q9BZS1
Gene Alias AIID/DIETER/IPEX/JM2/MGC141961/MGC141963/PIDX/XPID
Gene Symbols FOXP3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to smino acids 400-431 of human FOXP3.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50943
Test Specificity This antibody is specific to human FOXP3.
Target Species Human, Mouse
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.