Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FPGS, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89117273 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-273 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-273 Supplier Abnova Corporation Supplier No. PAB14801

Rabbit polyclonal antibody raised against synthetic peptide of FPGS.

This gene encodes the folylpolyglutamate synthetase enzyme. This enzyme has a central role in establishing and maintaining both cytosolic and mitochondrial folylpolyglutamate concentrations and, therefore, is essential for folate homeostasis and the survival of proliferating cells. This enzyme catalyzes the ATP-dependent addition of glutamate moieties to folate and folate derivatives. While several transcript variants may exist for this gene, the full-length natures of only two have been biologically validated to date. These two variants encode distinct isoforms. [provided by RefSeq

Specifications

Antigen FPGS
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Concentration Lot specific
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of FPGS.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.4 (1% BSA, 0.09% sodium azide)
Gene FPGS
Gene Symbols FPGS
Host Species Rabbit
Immunogen A synthetic peptide corresponding to internal region of human FPGS.
Purification Method Immunoaffinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2356
Test Specificity Molecular weight: 65 KDa. Epitope: Internal region.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.