Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FPR1 Rabbit anti-Chimpanzee, Gorilla, Human, Orangutan, Polyclonal Antibody, Abnova™

Catalog No. 89114105 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-105 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-105 Supplier Abnova Corporation Supplier No. PAB0284
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant FPR1.

Specifications

Antigen FPR1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant FPR1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% gelatin, 0.05% sodium azide)
Gene FPR1
Gene Alias FMLP/FPR
Gene Symbols FPR1
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 50-200 of human FPR1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2357
Target Species Chimpanzee, Gorilla, Human, Orangutan
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.