Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FREM1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89120801 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-120-801 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-120-801 Supplier Abnova Corporation Supplier No. PAB19270
Add to Cart
Add to Cart

Rabbit polyclonal antibody rasied against synthetic peptide of FREM1.

Specifications

Antigen FREM1
Applications ELISA, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody rasied against synthetic peptide of FREM1.
Dilution Western Blot (0.5-1 ug/mL) Immunocytochemistry (20 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene FREM1
Gene Accession No. NP_659403
Gene Alias C9orf143/C9orf145/C9orf154/FLJ25461/QBRICK/RP11-439N12.3
Gene Symbols FREM1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 15 amino acids near C-terminus of human FREM1.
Purification Method Peptide affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 158326
Target Species Human, Mouse
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.