Learn More
FRK, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of FRK.
Supplier: Abnova Corporation PAB3426
Description
The protein encoded by this gene belongs to the TYR family of protein kinases. This tyrosine kinase is a nuclear protein and may function during G1 and S phase of the cell cycle and suppress growth. [provided by RefSeq
Specifications
FRK | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of FRK. | |
In PBS (0.09% sodium azide) | |
P42685 | |
FRK | |
A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human FRK. | |
400 μL | |
Primary | |
Human | |
Liquid |
Flow Cytometry, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Flow Cytometry (1:10-50) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:10-50) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
FRK | |
GTK/PTK5/RAK | |
Rabbit | |
Protein G purification | |
RUO | |
2444 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only