Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FRS2 (phospho Y436), Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89130661 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-661 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-661 Supplier Abnova Corporation Supplier No. PAB29226
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic phosphopeptide of human FRS2.

Specifications

Antigen FRS2
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic phosphopeptide of human FRS2.
Dilution Immunohistochemistry (1:50-1:100) Western Blot (1:500-1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (without mg2+ and Ca2+), (50% glycerol, 0.02% sodium azide)
Gene FRS2
Gene Accession No. Q8WU20
Gene Alias FRS2A/FRS2alpha/SNT/SNT-1/SNT1
Gene Symbols FRS2
Host Species Rabbit
Immunogen Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding Y436 of human FRS2.
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10818
Test Specificity This antibody detects endogenous levels of FRS2 only when phosphorylated at tyrosine 436.
Target Species Human, Mouse
Content And Storage Store at -20°C.
Avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.