Learn More
FYN Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of FYN.
Supplier: Abnova Corporation PAB27162
Description
This gene is a member of the protein-tyrosine kinase oncogene family. It encodes a membrane-associated tyrosine kinase that has been implicated in the control of cell growth. The protein associates with the p85 subunit of phosphatidylinositol 3-kinase and interacts with the fyn-binding protein. Alternatively spliced transcript variants encoding distinct isoforms exist. [provided by RefSeq
Specifications
FYN | |
Polyclonal | |
Unconjugated | |
Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
FYN | |
MGC45350/SLK/SYN | |
Rabbit | |
Antigen affinity purification | |
RUO | |
2534 | |
Human, Mouse | |
Liquid |
Immunohistochemistry (PFA fixed), Western Blot | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of FYN. | |
In PBS, pH 7.2 (0.05% sodium azide) | |
P06241 | |
FYN | |
A synthetic peptide corresponding to human FYN. | |
100 μL | |
Primary | |
FYN polyclonal antibody detects endogenous levels of FYN protein. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only