Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-FZD2, Polyclonal, Abnova™

Catalog No. 89127892 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-127-892 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-127-892 Supplier Abnova Corporation Supplier No. PAB26251

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Specifications

Antigen FZD2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Formulation In PBS (0.09% sodium azide)
Gene Accession No. Q14332
Gene Symbols FZD2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 18 amino acid at extracellular domain of human FZD2.
Purification Method Affinity Purified
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) 2535
Target Species Bovine, Canine, Equine, Hamster, Human, Mouse, Pig, Rat
Content And Storage Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.